Gene Transcripts

Gene Symbol
Ankrd17
Entrez Gene ID
81702
Gene Name
ankyrin repeat domain 17
Gene Aliases
4933425K22Rik, A130069E23Rik, A930008M01, AA407558, AA516750, AU040470, Foe, Gtar, Mask, mKIAA0697
Chromosome Location
Chr. 5: 90227166 - 90367845 on Build GRCm38
UniGene ID
Mm.245522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030886.2 NP_112148.2 6 1167
NM_198010.2 NP_932127.2 6 1167
GenBank mRNA AF130371.1 - 5 708
AK044355.1 - 6 779
BC085166.1 - 6 685

View Full Product Details