Gene Transcripts

Gene Symbol
HSPA4
Entrez Gene ID
3308
Gene Name
heat shock protein family A (Hsp70) member 4
Gene Aliases
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY
Chromosome Location
Chr. 5: 133051970 - 133105017 on Build GRCh38
UniGene ID
Hs.90093
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002154.3 NP_002145.3 19 2776
GenBank mRNA AB023420.1 - 19 2773
AK295847.1 - 16 2161
AK297762.1 - 2 307
AK299710.1 - 8 1052
AK300050.1 - 15 2049
AK300770.1 - 18 2444
AK303382.1 - 13 1696
AK304578.1 - 15 1982
AK315862.1 - 5 642
AK315871.1 - 4 534
AK315875.1 - 3 457
AK315887.1 - 7 931
BC002526.2 - 5 619
BC110861.1 - 19 2636 6169048
BC126122.1 - 19 2737 8991838
BC126124.1 - 19 2737 8991840
BQ431980.1 - 6 712
BT007375.1 - 5 419
BU171424.1 - 1 132
BU686725.1 - 2 239
CF145856.1 - 2 300
FJ224293.1 - 19 2773

View Full Product Details