Gene Transcripts

Gene Symbol
ZNF655
Entrez Gene ID
79027
Gene Name
zinc finger protein 655
Gene Aliases
VIK, VIK-1
Chromosome Location
Chr. 7: 99557726 - 99576453 on Build GRCh38
UniGene ID
Hs.599798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009960.1 NP_001009960.1 3 2145
NM_001083956.1 NP_001077425.1 4 2423
NM_001085368.1 NP_001078837.1 4 2250
NM_138494.2 NP_612503.1 3 2318
XM_017012598.1 XP_016868087.1 6 3323
XM_017012599.1 XP_016868088.1 6 2481
XM_017012600.1 XP_016868089.1 6 2494
XM_017012601.1 XP_016868090.1 6 2480
XM_017012602.1 XP_016868091.1 5 3218
XM_017012603.1 XP_016868092.1 4 3343
XM_017012604.1 XP_016868093.1 4 2260
XM_017012605.1 XP_016868094.1 5 2271
XM_017012606.1 XP_016868095.1 3 2176
XM_017012607.1 XP_016868096.1 3 2162
XM_017012608.1 XP_016868097.1 6 3356
XM_017012609.1 XP_016868098.1 5 2370
XM_017012610.1 XP_016868099.1 5 2317
GenBank mRNA AK098231.1 - 1 24
AK122929.1 - 1 1402
AK128057.1 - 2 3382
AK316024.1 - 4 2241
AY099353.1 - 3 2144
BC011816.1 - 1 1413
BC024770.1 - 1 1664
BC037407.1 - 3 2108 5752559
CR749270.1 - 3 1926

View Full Product Details