Gene Transcripts

Gene Symbol
SUN1
Entrez Gene ID
23353
Gene Name
Sad1 and UNC84 domain containing 1
Gene Aliases
UNC84A
Chromosome Location
Chr. 7: 815557 - 874920 on Build GRCh38
UniGene ID
Hs.438072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130965.2 NP_001124437.1 16 1899
NM_001171944.1 NP_001165415.1 14 1649
NM_025154.5 NP_079430.3 15 1762
GenBank mRNA AB018353.1 - 17 1997
AB648918.1 - 18 2188
AF202724.1 - 7 798
AK022816.1 - 15 1754
AK025737.1 - 13 1424
AK074320.1 - 3 346
AK302896.1 - 14 1621
AK304150.1 - 12 1290
BC142707.1 - 16 1909 40148817
BC152418.1 - 16 1997 8862528
BX640866.1 - 7 1988
BX648893.1 - 7 1988
BX649069.1 - 14 1654

View Full Product Details