Gene Transcripts

Gene Symbol
HNRNPCL1
Entrez Gene ID
343069
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 1
Gene Aliases
HNRPCL1
Chromosome Location
Chr. 1: 12847377 - 12848756 on Build GRCh38
UniGene ID
Hs.502617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013631.2 NP_001013653.1 2 601
GenBank mRNA BC002696.2 - 2 570
BC137258.1 - 2 601 9021255
BC171866.1 - 1 439 9054520

Gene Symbol
HNRNPCL2
Entrez Gene ID
440563
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 2
Gene Aliases
HNRNPCP5
Chromosome Location
Chr. 1: 13115488 - 13116854 on Build GRCh38
UniGene ID
Hs.711869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136561.3 NP_001130033.3 2 587
GenBank mRNA BC144551.1 - 2 587
BC157858.1 - 1 439 9057077
BC171863.1 - 1 439 9054517

View Full Product Details