Gene Transcripts

Gene Symbol
Slc25a29
Entrez Gene ID
314441
Gene Name
solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29
Gene Aliases
Chromosome Location
Chr. 6: 132736968 - 132747762 on Build Rnor_6.0
UniGene ID
Rn.15047
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010958.1 NP_001010958.1 4 357
XM_006240541.2 XP_006240603.1 4 518
GenBank mRNA BC089065.1 - 4 357

View Full Product Details