Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
Chromosome Location
Chr. 2: 189674292 - 189763059 on Build GRCh38
UniGene ID
Hs.60772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144708.3 NP_653309.3 2 118
XM_011510673.2 XP_011508975.1 2 604
XM_011510674.1 XP_011508976.1 2 604
XM_011510675.2 XP_011508977.1 2 604
XM_011510676.2 XP_011508978.1 2 604
XM_011510677.2 XP_011508979.1 2 604
XM_011510679.2 XP_011508981.1 2 604
XM_011510680.1 XP_011508982.1 2 604
XM_011510681.1 XP_011508983.1 2 604
XM_011510682.2 XP_011508984.1 2 604
XM_011510686.2 XP_011508988.1 2 604
XM_011510688.1 XP_011508990.1 2 604
XM_011510689.2 XP_011508991.1 2 604
XM_017003413.1 XP_016858902.1 2 604
XM_017003414.1 XP_016858903.1 2 604
XM_017003417.1 XP_016858906.1 2 604

View Full Product Details