Gene Transcripts

Gene Symbol
C2orf71
Entrez Gene ID
388939
Gene Name
chromosome 2 open reading frame 71
Gene Aliases
RP54
Chromosome Location
Chr. 2: 29061690 - 29074261 on Build GRCh38
UniGene ID
Hs.354243
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029883.2 NP_001025054.1 2 4662
GenBank mRNA AK092250.1 - 2 1197

View Full Product Details