Gene Transcripts

Gene Symbol
Olfr1513
Entrez Gene ID
258008
Gene Name
olfactory receptor 1513
Gene Aliases
MOR223-10, MOR223-7P
Chromosome Location
Chr. 14: 52349103 - 52350044 on Build GRCm38
UniGene ID
Mm.377327
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012269.2 NP_001012269.2 1 340

View Full Product Details