Gene Transcripts

Gene Symbol
Olfr57
Entrez Gene ID
18357
Gene Name
olfactory receptor 57
Gene Aliases
IF12, MOR139-3
Chromosome Location
Chr. 10: 79034723 - 79035802 on Build GRCm38
UniGene ID
Mm.246544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147041.2 NP_667252.1 1 1018
GenBank mRNA BC141307.1 - 1 1022

View Full Product Details