Gene Transcripts

Gene Symbol
HNRNPCL1
Entrez Gene ID
343069
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 1
Gene Aliases
HNRPCL1
Chromosome Location
Chr. 1: 12847377 - 12848756 on Build GRCh38
UniGene ID
Hs.502617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013631.2 NP_001013653.1 2 495
GenBank mRNA BC002696.2 - 2 464
BC137258.1 - 2 495 9021255
BC171866.1 - 1 333 9054520
BX283835.1 - 2 465

Gene Symbol
HNRNPCL3
Entrez Gene ID
649330
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 3
Gene Aliases
HNRNPCL2
Chromosome Location
Chr. 1: 13061232 - 13062237 on Build GRCh38
UniGene ID
Hs.502617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146181.2 NP_001139653.1 1 331
GenBank mRNA BC171859.1 - 1 333
BC171865.1 - 1 333 9054519

Gene Symbol
HNRNPCL4
Entrez Gene ID
101060301
Gene Name
heterogeneous nuclear ribonucleoprotein C-like 4
Gene Aliases
Chromosome Location
Chr. 1: 13164555 - 13165482 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302551.1 NP_001289480.1 1 253

View Full Product Details