Gene Transcripts

Gene Symbol
FAM167B
Entrez Gene ID
84734
Gene Name
family with sequence similarity 167 member B
Gene Aliases
C1orf90
Chromosome Location
Chr. 1: 32247217 - 32248860 on Build GRCh38
UniGene ID
Hs.18449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032648.2 NP_116037.2 2 821
GenBank mRNA BC004269.1 - 2 805
BX091465.1 - 1 53

View Full Product Details