Gene Transcripts

Gene Symbol
GLMP
Entrez Gene ID
112770
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1orf85, NCU-G1
Chromosome Location
Chr. 1: 156292687 - 156295689 on Build GRCh38
UniGene ID
Hs.202522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256604.1 NP_001243533.1 2 391
NM_001256608.1 NP_001243537.1 2 391
NM_001256609.1 NP_001243538.1 2 620
NM_144580.2 NP_653181.1 2 391
XM_017000149.1 XP_016855638.1 2 387
GenBank mRNA AK075349.1 - 2 360
AK296157.1 - 2 588
AY358450.1 - 2 364
BC011575.1 - 2 374
BC018757.1 - 2 360 4850051
BC036340.1 - 2 307
CX756681.1 - 2 306
DA537642.1 - 2 391

View Full Product Details