Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153712.4 | NP_714923.1 | 3 | 558 | |
XM_005263599.3 | XP_005263656.1 | 3 | 597 | ||
XM_011510665.2 | XP_011508967.1 | 3 | 597 | ||
GenBank mRNA | AB071393.2 | - | 3 | 403 | |
AK126549.1 | - | 1 | 50 | ||
BC036819.1 | - | 3 | 570 | 5767758 |