Gene Transcripts

Gene Symbol
IFI27L1
Entrez Gene ID
122509
Gene Name
interferon alpha inducible protein 27 like 1
Gene Aliases
FAM14B, ISG12C
Chromosome Location
Chr. 14: 94081293 - 94102714 on Build GRCh38
UniGene ID
Hs.19414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145249.2 NP_660292.1 4 404
NM_206949.2 NP_996832.1 4 358
XM_005267314.4 XP_005267371.1 3 2949
XM_011536405.2 XP_011534707.1 4 374
XM_011536406.1 XP_011534708.1 5 506
XM_011536407.1 XP_011534709.1 5 509
XM_017020961.1 XP_016876450.1 4 255
XM_017020962.1 XP_016876451.1 3 224
XM_017020963.1 XP_016876452.1 4 257
GenBank mRNA BC015423.1 - 4 368

View Full Product Details