Gene Transcripts
- Gene Symbol
- RPS6KB2
- Entrez Gene ID
- 6199
- Gene Name
- ribosomal protein S6 kinase B2
- Gene Aliases
- KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
- Chromosome Location
- Chr. 11: 67428460 - 67435408 on Build GRCh38
- UniGene ID
- Hs.534345
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003952.2 |
NP_003943.2
|
3, 4 |
330 |
|
XM_005274164.1 |
XP_005274221.1
|
2, 3 |
278 |
|
XM_005274165.4 |
XP_005274222.1
|
3, 4 |
303 |
|
XM_006718655.3 |
XP_006718718.1
|
3, 4 |
311 |
|
XM_006718656.3 |
XP_006718719.1
|
2, 3 |
262 |
|
XM_006718657.1 |
XP_006718720.1
|
2, 3 |
244 |
|
XM_017018108.1 |
XP_016873597.1
|
3, 4 |
325 |
|
GenBank mRNA |
AB016869.1 |
- |
3, 4 |
302 |
|
AB019245.1 |
- |
3, 4 |
300 |
|
AB208827.1 |
- |
1, 2 |
744 |
|
AF076931.1 |
- |
3, 4 |
283 |
|
AF099739.1 |
- |
3, 4 |
287 |
|
AK297527.1 |
- |
3, 4 |
296 |
|
BC000094.2 |
- |
3, 4 |
303 |
3508140 |
BC006106.1 |
- |
3, 4 |
286 |
2959036 |
BC136564.1 |
- |
3, 4 |
304 |
9020554 |
View Full Product Details