Gene Transcripts

Gene Symbol
Ceacam3
Entrez Gene ID
24926
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 3
Gene Aliases
CGM4AA, Cea, Ceacam5, Cear, Cgm1, Cgm4, RATCEAA, RATCGM4AA
Chromosome Location
Chr. 1: 79122593 - 79135127 on Build Rnor_6.0
UniGene ID
Rn.10064
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012702.2 NP_036834.2 3 738
GenBank mRNA BC098921.1 - 3 738
M32474.1 - 3 723

View Full Product Details