Gene Transcripts
- Gene Symbol
- Itgam
- Entrez Gene ID
- 25021
- Gene Name
- integrin, alpha M
- Gene Aliases
- Cd11b
- Chromosome Location
- Chr. 1: 199495307 - 199545738 on Build Rnor_6.0
- UniGene ID
- Rn.54465
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012711.1 |
NP_036843.1
|
10 |
1160 |
|
GenBank mRNA |
AF268593.1 |
- |
10 |
1160 |
|
View Full Product Details