Gene Transcripts

Gene Symbol
Ptpn11
Entrez Gene ID
25622
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
Shp2
Chromosome Location
Chr. 12: 40895515 - 40955999 on Build Rnor_6.0
UniGene ID
Rn.98209
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177593.1 NP_001171064.1 3, 4 516
NM_013088.2 NP_037220.2 3, 4 516
GenBank mRNA U09307.1 - 3, 4 441

View Full Product Details