Gene Transcripts

Gene Symbol
Brd1
Entrez Gene ID
223770
Gene Name
bromodomain containing 1
Gene Aliases
1110059H06Rik, AI316859, mKIAA4191
Chromosome Location
Chr. 15: 88687034 - 88734226 on Build GRCm38
UniGene ID
Mm.254438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033274.3 NP_001028446.2 2 1389
GenBank mRNA AK149714.1 - 2 1401
AK220233.1 - 1 310

View Full Product Details