Gene Transcripts

Gene Symbol
Cfap221
Entrez Gene ID
226356
Gene Name
cilia and flagella associated protein 221
Gene Aliases
Gm101, Pcdp1
Chromosome Location
Chr. 1: 119923340 - 119997471 on Build GRCm38
UniGene ID
Mm.297290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115074.1 NP_001108546.1 14 1497
GenBank mRNA EF632061.1 - 14 1497

View Full Product Details