Gene Transcripts

Gene Symbol
Cyp11a1
Entrez Gene ID
29680
Gene Name
cytochrome P450, family 11, subfamily a, polypeptide 1
Gene Aliases
Cyp11a, Cypxia1, P450scc
Chromosome Location
Chr. 8: 62798342 - 62809674 on Build Rnor_6.0
UniGene ID
Rn.1401
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017286.2 NP_058982.1 3 519
GenBank mRNA BC089100.1 - 3 544
DQ515795.1 - 3 506
J05156.1 - 3 539

View Full Product Details