Gene Transcripts

Gene Symbol
Esp1
Entrez Gene ID
619517
Gene Name
exocrine gland secreted peptide 1
Gene Aliases
EG619517, Gm6084
Chromosome Location
Chr. 17: 40727120 - 40731782 on Build GRCm38
UniGene ID
Mm.391018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038500.2 NP_001033589.1 3 299
GenBank mRNA AB194091.1 - 3 328

View Full Product Details