Gene Transcripts

Gene Symbol
Mkrn2os
Entrez Gene ID
70291
Gene Name
makorin, ring finger protein 2, opposite strand
Gene Aliases
2510049J12Rik
Chromosome Location
Chr. 6: 115583547 - 115592576 on Build GRCm38
UniGene ID
Mm.133694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101431.1 NP_001094901.1 4 1435
GenBank mRNA AK028760.1 - 3 1308

View Full Product Details