Gene Transcripts

Gene Symbol
Impg1
Entrez Gene ID
66014
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
Mlgapc
Chromosome Location
Chr. 8: 87733545 - 87879888 on Build Rnor_6.0
UniGene ID
Rn.64488
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023958.1 NP_076448.1 2 391
GenBank mRNA AB047843.1 - 2 391

View Full Product Details