Gene Transcripts
- Gene Symbol
- Impg1
- Entrez Gene ID
- 66014
- Gene Name
- interphotoreceptor matrix proteoglycan 1
- Gene Aliases
- Mlgapc
- Chromosome Location
- Chr. 8: 87733545 - 87879888 on Build Rnor_6.0
- UniGene ID
- Rn.64488
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023958.1 |
NP_076448.1
|
16, 17 |
2453 |
|
GenBank mRNA |
AB047843.1 |
- |
16, 17 |
2453 |
|
View Full Product Details