Gene Transcripts

Gene Symbol
Tas2r105
Entrez Gene ID
78985
Gene Name
taste receptor, type 2, member 105
Gene Aliases
T2R9
Chromosome Location
Chr. 4: 165827885 - 165828814 on Build Rnor_6.0
UniGene ID
Rn.48788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023999.1 NP_076489.1 1 884
GenBank mRNA AF227146.1 - 1 884

View Full Product Details