Gene Transcripts

Gene Symbol
Magt1
Entrez Gene ID
116967
Gene Name
magnesium transporter 1
Gene Aliases
Iag2
Chromosome Location
Chr. X: 77023423 - 77061603 on Build Rnor_6.0
UniGene ID
Rn.43578
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053946.1 NP_446398.1 2 179
GenBank mRNA AF008554.2 - 2 179

View Full Product Details