Gene Transcripts
- Gene Symbol
- Galnt10
- Entrez Gene ID
- 170501
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 10
- Gene Aliases
- Galnt9
- Chromosome Location
- Chr. 10: 43067316 - 43208992 on Build Rnor_6.0
- UniGene ID
- Rn.60224
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130742.1 |
NP_570098.1
|
1, 2 |
158 |
|
GenBank mRNA |
AF241241.1 |
- |
1, 2 |
158 |
|
View Full Product Details