Gene Transcripts
- Gene Symbol
- Ptpn21
- Entrez Gene ID
- 171070
- Gene Name
- protein tyrosine phosphatase, non-receptor type 21
- Gene Aliases
- Ptp2E
- Chromosome Location
- Chr. 6: 122656500 - 122721496 on Build Rnor_6.0
- UniGene ID
- Rn.11310
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133545.1 |
NP_598229.1
|
4 |
651 |
|
GenBank mRNA |
U17971.1 |
- |
4 |
651 |
|
View Full Product Details