Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007395.3 | NP_031421.1 | 7, 8 | 1289 | |
GenBank mRNA | AK154578.1 | - | 1312 | ||
AK157910.1 | - | 7, 8 | 1290 | ||
BC059832.1 | - | 1282 | |||
BC145775.1 | - | 7, 8 | 1342 | 40131078 | |
BC145777.1 | - | 7, 8 | 1342 | 40131080 | |
Z31663.1 | - | 7, 8 | 1278 |