Gene Transcripts

Gene Symbol
Klra2
Entrez Gene ID
16633
Gene Name
killer cell lectin-like receptor, subfamily A, member 2
Gene Aliases
Klra30, Ly49b
Chromosome Location
Chr. 6: 131198976 - 131247723 on Build GRCm38
UniGene ID
Mm.4783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170851.1 NP_001164322.1 4 408
NM_008462.5 NP_032488.4 4 408
GenBank mRNA AF253057.1 - 3 242
AF253058.1 - 3 242
AF253059.1 - 3 242
AF395446.1 - 3 242
AK017140.1 - 4 399
AY860971.1 - 3 242
BC064711.1 - 4 408 30295755
EF025059.1 - 3 242
U10304.1 - 4 336

View Full Product Details