Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166623.1 | NP_001160095.1 | 3 | 342 | |
NM_011252.4 | NP_035382.1 | 3 | 339 | ||
GenBank mRNA | AJ237846.1 | - | 3 | 269 | |
AJ237847.1 | - | 3 | 284 | ||
AK083899.1 | - | 3 | 339 | ||
AK135164.1 | - | 3 | 344 | ||
AK165006.1 | - | 3 | 285 | ||
AK207357.1 | - | 3 | 203 | ||
BC003710.1 | - | 3 | 337 | 3599678 | |
BY279114.1 | - | 3 | 339 | ||
CK619886.1 | - | 2 | 145 |