Gene Transcripts

Gene Symbol
Sun1
Entrez Gene ID
77053
Gene Name
Sad1 and UNC84 domain containing 1
Gene Aliases
4632417G13Rik, 5730434D03Rik, Unc84a, mKIAA0810
Chromosome Location
Chr. 5: 139200558 - 139249839 on Build GRCm38
UniGene ID
Mm.210845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256115.1 NP_001243044.1 5, 6 572
NM_001256116.1 NP_001243045.1 4, 5 509
NM_001256117.1 NP_001243046.1 5, 6 572
NM_001256118.1 NP_001243047.1 4, 5 509
NM_024451.2 NP_077771.1 5, 6 572
GenBank mRNA AK014585.1 - 5, 6 570
AK036187.1 - 5, 6 549
AK160281.1 - 5, 6 530
AK161959.1 - 5, 6 544
AK162533.1 - 5, 6 543
AK167686.1 - 5, 6 544
AK181636.1 - 2, 3 96
AK194327.1 - 2, 3 95
AK208681.1 - 3, 4 223
AK210229.1 - 2, 3 230
AY682989.1 - 4, 5 489
BC047928.1 - 5, 6 539 5321879
BC048156.1 - 5, 6 541 5698402
BQ551727.1 - 4, 5 513
HQ402597.1 - 4, 5 489

View Full Product Details