Gene Transcripts

Gene Symbol
Utp11l
Entrez Gene ID
67205
Gene Name
UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Gene Aliases
2700082D03Rik, CGI-94, Cgi94
Chromosome Location
Chr. 4: 124678232 - 124693612 on Build GRCm38
UniGene ID
Mm.259121
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026031.3 NP_080307.1 4, 5 396
GenBank mRNA AK007361.1 - 2, 3 167
AK012550.1 - 4, 5 397
AK145266.1 - 4, 5 429
AK147113.1 - 4, 5 427
BC061074.1 - 4, 5 425 6706808

View Full Product Details