Gene Transcripts

Gene Symbol
Cfap52
Entrez Gene ID
71860
Gene Name
cilia and flagella associated protein 52
Gene Aliases
1700019F09Rik, 4933417B11Rik, WDRPUH, Wdr16
Chromosome Location
Chr. 11: 67924806 - 67965929 on Build GRCm38
UniGene ID
Mm.441823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027963.2 NP_082239.2 2 167
GenBank mRNA AK006118.1 - 2 167
AK016839.1 - 2 178
BC118541.1 - 2 166 40058756

View Full Product Details