Gene Transcripts

Gene Symbol
Polr3h
Entrez Gene ID
78929
Gene Name
polymerase (RNA) III (DNA directed) polypeptide H
Gene Aliases
5031409G22Rik, RPC8
Chromosome Location
Chr. 15: 81915030 - 81926247 on Build GRCm38
UniGene ID
Mm.215115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030229.4 NP_084505.2 6 2410
GenBank mRNA AK019868.2 - 6 2411

View Full Product Details