Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167763.1 | NP_001161235.1 | 3 | 299 | |
NM_031177.4 | NP_112454.2 | 3 | 299 | ||
GenBank mRNA | AK031308.1 | - | 3 | 187 | |
AK044456.1 | - | 3 | 299 | ||
AK131115.1 | - | 3 | 226 | ||
AK167330.1 | - | 3 | 254 | ||
AK167412.1 | - | 3 | 254 | ||
AK199875.1 | - | 2 | 128 | ||
AK206333.1 | - | 2 | 127 | ||
AK218729.1 | - | 1 | 60 | ||
BC060621.1 | - | 3 | 247 | ||
BC066083.1 | - | 3 | 218 | 30355922 | |
BC067415.1 | - | 3 | 215 | 30063124 |