Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001169153.1 | NP_001162624.1 | 6 | 935 | |
NM_145634.3 | NP_663609.2 | 6 | 914 | ||
GenBank mRNA | AB292061.1 | - | 6 | 824 | |
AF251703.1 | - | 5 | 727 | ||
AK089313.1 | - | 6 | 935 | ||
AK154273.1 | - | 5 | 745 | ||
AK155367.1 | - | 6 | 912 | ||
AY214460.1 | - | 6 | 866 | ||
AY457047.1 | - | 6 | 824 | ||
BC057864.1 | - | 6 | 879 | 5318856 | |
JX073136.1 | - | 5 | 780 |