Gene Transcripts

Gene Symbol
Olfr109
Entrez Gene ID
258832
Gene Name
olfactory receptor 109
Gene Aliases
MOR250-1
Chromosome Location
Chr. 17: 37466208 - 37467152 on Build GRCm38
UniGene ID
Mm.377712
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146835.1 NP_667046.1 1 889
GenBank mRNA BC130246.1 - 1 893

View Full Product Details