Gene Transcripts

Gene Symbol
Olfr1228
Entrez Gene ID
258973
Gene Name
olfactory receptor 1228
Gene Aliases
MOR233-1
Chromosome Location
Chr. 2: 89248721 - 89253707 on Build GRCm38
UniGene ID
Mm.463734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146971.1 NP_667182.1 1 374
GenBank mRNA BC125451.1 - 1 457
BC125453.1 - 1 457 40129968

View Full Product Details