Gene Transcripts

Gene Symbol
Olfr449
Entrez Gene ID
259067
Gene Name
olfactory receptor 449
Gene Aliases
MOR103-1
Chromosome Location
Chr. 6: 42834384 - 42838882 on Build GRCm38
UniGene ID
Mm.451673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147064.1 NP_667275.1 1 880
GenBank mRNA BC138166.1 - 1 911

View Full Product Details