Gene Transcripts

Gene Symbol
Mamstr
Entrez Gene ID
74490
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
2810022D01Rik, 5430432N15Rik, AW743872, Mastr
Chromosome Location
Chr. 7: 45639200 - 45648206 on Build GRCm38
UniGene ID
Mm.107442
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172418.2 NP_766006.2 8 850
GenBank mRNA AK017382.1 - 7 1158
AK036109.1 - 5 968
AK052976.1 - 2 2003
BC107175.1 - 3 369
BC107176.1 - 3 371
BC119481.1 - 3 369
BC119482.1 - 3 369
BC127227.1 - 3 371
BC127228.1 - 3 371
DQ534901.1 - 8 834

View Full Product Details