Gene Transcripts

Gene Symbol
Tprg
Entrez Gene ID
71338
Gene Name
transformation related protein 63 regulated
Gene Aliases
5430420C16Rik, Tprg1
Chromosome Location
Chr. 16: 25286817 - 25423588 on Build GRCm38
UniGene ID
Mm.126851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175165.3 NP_780374.1 2 354
GenBank mRNA AK017323.1 - 2 350
AK036796.1 - 2 357

View Full Product Details