Gene Transcripts

Gene Symbol
Nlrp2
Entrez Gene ID
232827
Gene Name
NLR family, pyrin domain containing 2
Gene Aliases
E330007A02Rik, NBS1, Nalp2, PAN1, PYPAF2
Chromosome Location
Chr. 7: 5298547 - 5351035 on Build GRCm38
UniGene ID
Mm.246598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177690.3 NP_808358.2 2 313
GenBank mRNA AK054264.1 - 2 314
AY360473.1 - 2 314
DQ066881.1 - 2 314

View Full Product Details