Gene Transcripts

Gene Symbol
Sertm1
Entrez Gene ID
329641
Gene Name
serine rich and transmembrane domain containing 1
Gene Aliases
6030405A18Rik
Chromosome Location
Chr. 3: 54897068 - 54915887 on Build GRCm38
UniGene ID
Mm.51187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177854.4 NP_808522.1 3 832
GenBank mRNA AK031306.1 - 3 833
BC058995.1 - 2 645 5708109

View Full Product Details