Gene Transcripts

Gene Symbol
Zer1
Entrez Gene ID
227693
Gene Name
zyg-11 related, cell cycle regulator
Gene Aliases
C230075L19Rik, Zyg11bl
Chromosome Location
Chr. 2: 30097283 - 30124804 on Build GRCm38
UniGene ID
Mm.44526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290503.1 NP_001277432.1 4 1085
NM_178694.4 NP_848809.2 4 1085
GenBank mRNA AK029290.1 - 4 1059
AK032153.1 - 4 1055
AK032318.1 - 3 731
AK035712.1 - 1 37
AK048853.1 - 4 980
AK081355.1 - 4 962
AK087046.1 - 4 958
AK155401.1 - 4 1032
BC048924.1 - 4 928 6312691
BC057021.1 - 4 1029 6825937

View Full Product Details