Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
Chromosome Location
Chr. 16: 72008744 - 72027659 on Build GRCh38
UniGene ID
Hs.654427
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001361.4 NP_001352.2 3, 4 445
XM_005255827.3 XP_005255884.1 3, 4 750
XM_017022990.1 XP_016878479.1 3, 4 525
GenBank mRNA AK292293.1 - 4, 5 601
AU130529.1 - 4, 5 445
BC065245.1 - 3, 4 425 6064723
BI253747.1 - 3, 4 429
BM795077.1 - 2, 3 220
M94065.1 - 4, 5 427

View Full Product Details