Gene Transcripts

Gene Symbol
ONECUT1
Entrez Gene ID
3175
Gene Name
one cut homeobox 1
Gene Aliases
HNF-6, HNF6, HNF6A
Chromosome Location
Chr. 15: 52756963 - 52790264 on Build GRCh38
UniGene ID
Hs.658573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004498.2 NP_004489.1 1 1138
XM_011521487.2 XP_011519789.1 1 1390
GenBank mRNA BC140830.1 - 1 1105
EU532019.1 - 1 758
U77975.1 - 1 491
U96173.1 - 1 1138

View Full Product Details