Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193493.1 | NP_001180422.1 | 7 | 853 | |
NM_003038.4 | NP_003029.2 | 8 | 1721 | ||
XM_006712079.2 | XP_006712142.1 | 8 | 1087 | ||
GenBank mRNA | AB026689.1 | - | 8 | 1459 | |
AK126636.1 | - | 7 | 2911 | ||
AK223486.1 | - | 8 | 1683 | ||
AK295687.1 | - | 7 | 808 | ||
AK298472.1 | - | 9 | 1632 | ||
AK313052.1 | - | 8 | 1685 | ||
AK315986.1 | - | 9 | 1643 | ||
BC026216.1 | - | 8 | 1622 | 4420405 | |
BC072423.1 | - | 8 | 1681 | 6186177 | |
L14595.1 | - | 8 | 1621 | ||
L19444.1 | - | 8 | 1600 |